ENA018109710 Enamine

4-(3,4-dimethylphenyl)-1-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD04621704
  • Cas Number : 730951-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06598
Availability details for
Quantity
  • Availability
  • Add to Cart