ENA018109711 Enamine

2,5-dimethyl-1-(2-methylpropyl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04621551
  • Cas Number : 730950-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06599
Availability details for
Quantity
  • Availability
  • Add to Cart