ENA018109730 Enamine

2-({5-[(4-ethylphenyl)amino]-1,3,4-thiadiazol-2-yl}sulfanyl)acetic acid

  • MDL number MFCD04612074
  • Cas Number : 571919-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-228
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06621
Availability details for
Quantity
  • Availability
  • Add to Cart