ENA018109737 Enamine

N-(4-hydroxyphenyl)furan-2-carboxamide

  • MDL number MFCD00033635
  • Cas Number : 4104-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06629
Availability details for
Quantity
  • Availability
  • Add to Cart