ENA018109738 Enamine

2-chloro-N-(1-cyano-1,2-dimethylpropyl)acetamide

  • MDL number MFCD04621514
  • Cas Number : 730950-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06630
Availability details for
Quantity
  • Availability
  • Add to Cart