ENA018109740 Enamine

3-[(4-tert-butylphenyl)formamido]propanoic acid

  • MDL number MFCD02936746
  • Cas Number : 111830-21-8
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06632
Availability details for
Quantity
  • Availability
  • Add to Cart