ENA018109751 Enamine

N2-(4-fluorophenyl)-1,3,5-triazine-2,4-diamine

  • MDL number MFCD01320741
  • Cas Number : 1549-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06644
Availability details for
Quantity
  • Availability
  • Add to Cart