ENA018109752 Enamine

N-o-Tolyl-[1,3,5]triazine-2,4-diamine

  • MDL number MFCD00547705
  • Cas Number : 58759-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06645

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06645
Availability details for
Quantity
  • Availability
  • Add to Cart