ENA018109756 Enamine

3-[1-(2,3-dimethoxyphenyl)-2-nitroethyl]-1H-indole

  • MDL number MFCD04612892
  • Cas Number : 33723-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06650
Availability details for
Quantity
  • Availability
  • Add to Cart