ENA018109777 Enamine

N-(2-chloroacetyl)-1-methyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD04621544
  • Cas Number : 730950-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06674
Availability details for
Quantity
  • Availability
  • Add to Cart