ENA018109786 Enamine

N-(5-chloro-2-methoxyphenyl)-3-oxobutanamide

  • MDL number MFCD00026252
  • Cas Number : 52793-11-0
  • Molecular Weight : 241.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06684
Availability details for
Quantity
  • Availability
  • Add to Cart