ENA018109788 Enamine

4-ethyl-7-hydroxy-2H-chromen-2-one

  • MDL number MFCD00276926
  • Cas Number : 64231-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-129
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06686
Availability details for
Quantity
  • Availability
  • Add to Cart