ENA018109790 Enamine

4-Isobutoxy-3-methoxy-benzaldehyde oxime

  • MDL number MFCD04612958
  • Cas Number : 556020-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06689

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06689
Availability details for
Quantity
  • Availability
  • Add to Cart