ENA018109798 Enamine

7-hydroxy-4-(propan-2-yl)-2H-chromen-2-one

  • MDL number MFCD04612954
  • Cas Number : 23251-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06697
Availability details for
Quantity
  • Availability
  • Add to Cart