ENA018109799 Enamine

3-(methoxymethyl)-1-benzofuran-2-carboxylic acid

  • MDL number MFCD04613029
  • Cas Number : 734546-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06698
Availability details for
Quantity
  • Availability
  • Add to Cart