ENA018109804 Enamine

5-Isobutyl-5-phenyl-imidazolidine-2,4-dione

  • MDL number MFCD04613018
  • Cas Number : 100615-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06704

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06704
Availability details for
Quantity
  • Availability
  • Add to Cart