ENA018109809 Enamine

5-(2-fluorophenyl)-4-(prop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04612962
  • Cas Number : 694461-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06710
Availability details for
Quantity
  • Availability
  • Add to Cart