ENA018109813 Enamine

5-amino-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD04614621
  • Cas Number : 88508-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06715
Availability details for
Quantity
  • Availability
  • Add to Cart