ENA018109817 Enamine

N-(3-Chloro-phenyl)-2-(3-oxo-1,2,3,4-tetrahydro-quinoxalin-2-yl)-acetamide

  • MDL number MFCD01183986
  • Cas Number : 318235-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06719

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06719
Availability details for
Quantity
  • Availability
  • Add to Cart