ENA018109851 Enamine

5-Chlorosulfonyl-isophthalic acid dimethyl ester

  • MDL number MFCD04613028
  • Cas Number : 29710-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06759

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06759
Availability details for
Quantity
  • Availability
  • Add to Cart