ENA018109867 Enamine

2-amino-2-cyclopropylpropanenitrile

  • MDL number MFCD04621701
  • Cas Number : 37024-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06776

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-06776
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart