ENA018109869 Enamine

3-(hydroxyimino)-5,7-dimethyl-2,3-dihydro-1H-indol-2-one

  • Cas Number : 324029-69-8
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06779
Availability details for
Quantity
  • Availability
  • Add to Cart