ENA018109886 Enamine

N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]-2-chloro-N-(tetrahydrofuran-2-ylmethyl)acetamide

  • MDL number MFCD04614714
  • Cas Number : 735322-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06801
Availability details for
Quantity
  • Availability
  • Add to Cart