ENA018109887 Enamine

2-(chloromethyl)-5-thien-2-yl-1,3,4-oxadiazole

  • MDL number MFCD01885819
  • Cas Number : 727374-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06802

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06802
Availability details for
Quantity
  • Availability
  • Add to Cart