ENA018109890 Enamine

5-chloro-1-[(2-chlorophenyl)methyl]-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04624245
  • Cas Number : 956374-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06805
Availability details for
Quantity
  • Availability
  • Add to Cart