ENA018109891 Enamine

8-(chloromethyl)-6-fluoro-2-phenyl-2,4-dihydro-1,3-benzodioxine

  • MDL number MFCD04624266
  • Cas Number : 730992-77-5
  • Molecular Weight : 278.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06806
Availability details for
Quantity
  • Availability
  • Add to Cart