ENA018109894 Enamine

2-(2-chlorophenyl)-2-(1H-indol-3-yl)ethan-1-amine

  • MDL number MFCD04614721
  • Cas Number : 735322-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06809
Availability details for
Quantity
  • Availability
  • Add to Cart