ENA018109901 Enamine

3-amino-N,N-diethyl-4-(3-methylphenoxy)benzene-1-sulfonamide

  • MDL number MFCD04614728
  • Cas Number : 735322-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06816
Availability details for
Quantity
  • Availability
  • Add to Cart