ENA018109906 Enamine

3-methyl-4-oxo-4,6,7,8-tetrahydropyrrolo[1,2-a]thieno[2,3-d]pyrimidine-2-carboxylic acid

  • MDL number MFCD04624240
  • Cas Number : 730992-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241...243
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06821

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06821
Availability details for
Quantity
  • Availability
  • Add to Cart