ENA018109907 Enamine

6-(trifluoromethyl)-1H,2H,3H,9H-pyrrolo[2,1-b]quinazolin-9-one

  • MDL number MFCD04624248
  • Cas Number : 730992-47-9
  • Molecular Weight : 254.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06822
Availability details for
Quantity
  • Availability
  • Add to Cart