ENA018109908 Enamine

1H,5H,6H,10bH-pyrazolo[1,5-c]quinazoline-5-thione

  • MDL number MFCD02948845
  • Cas Number : 59553-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06823
Availability details for
Quantity
  • Availability
  • Add to Cart