ENA018109909 Enamine

4-methyl-2-(piperidin-1-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD04624838
  • Cas Number : 730997-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06824
Availability details for
Quantity
  • Availability
  • Add to Cart