ENA018109929 Enamine

2-chloro-N-(1,1-dioxo-1lambda6-thiolan-3-yl)-N-propylacetamide

  • MDL number MFCD04617228
  • Cas Number : 727694-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06848
Availability details for
Quantity
  • Availability
  • Add to Cart