ENA018109958 Enamine

3-(cyclopentylsulfamoyl)-4-methoxybenzoic acid

  • MDL number MFCD04614079
  • Cas Number : 735305-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-137
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06881
Availability details for
Quantity
  • Availability
  • Add to Cart