ENA018109964 Enamine

[(4-amino-3-methylphenyl)sulfanyl]formonitrile

  • MDL number MFCD00462863
  • Cas Number : 33192-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06887
Availability details for
Quantity
  • Availability
  • Add to Cart