ENA018109971 Enamine

2-(3,3-dimethyl-2-oxobutylidene)-3-methyl-1,3-thiazolidin-4-one

  • MDL number MFCD04615275
  • Cas Number : 735342-66-2
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06894
Availability details for
Quantity
  • Availability
  • Add to Cart