ENA018109984 Enamine

[(4-amino-3,5-dimethylphenyl)sulfanyl]formonitrile

  • MDL number MFCD00649562
  • Cas Number : 14031-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06908
Availability details for
Quantity
  • Availability
  • Add to Cart