ENA018109986 Enamine

ethyl 2-amino-4-methyl-5-phenylthiophene-3-carboxylate

  • MDL number MFCD00652592
  • Cas Number : 4815-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06910
Availability details for
Quantity
  • Availability
  • Add to Cart