ENA018109991 Enamine

2-(chloromethyl)-3-(2-oxopropyl)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04624267
  • Cas Number : 730992-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-254
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06915
Availability details for
Quantity
  • Availability
  • Add to Cart