ENA018109992 Enamine

2-(furan-2-carbonyl)-1-benzofuran-3-amine

  • MDL number MFCD04623493
  • Cas Number : 730976-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06916
Availability details for
Quantity
  • Availability
  • Add to Cart