ENA018110005 Enamine

5-amino-3-(5-aminopentyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD02370774
  • Cas Number : 113513-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06931
Availability details for
Quantity
  • Availability
  • Add to Cart