ENA018110006 Enamine

5-amino-3-(3-aminopropyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD02370767
  • Cas Number : 113513-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06932
Availability details for
Quantity
  • Availability
  • Add to Cart