ENA018110007 Enamine

4-methyl-2-(morpholin-4-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD04624258
  • Cas Number : 90437-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06933
Availability details for
Quantity
  • Availability
  • Add to Cart