ENA018110015 Enamine

4-nitro-N1-[3-(trifluoromethyl)phenyl]benzene-1,2-diamine

  • MDL number MFCD04614041
  • Cas Number : 735305-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06942
Availability details for
Quantity
  • Availability
  • Add to Cart