ENA018110061 Enamine

5-{[(2-chlorophenyl)methyl]sulfanyl}-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04626533
  • Cas Number : 731012-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06993
Availability details for
Quantity
  • Availability
  • Add to Cart