ENA018110069 Enamine

3-(diethylsulfamoyl)-4-methylbenzoic acid

  • MDL number MFCD00625728
  • Cas Number : 313259-91-5
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07001
Availability details for
Quantity
  • Availability
  • Add to Cart