ENA018110079 Enamine

4-(4-chlorophenyl)-5-(4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02246701
  • Cas Number : 142142-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07014
Availability details for
Quantity
  • Availability
  • Add to Cart