ENA018110080 Enamine

4-(4-methylphenyl)-5-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01046343
  • Cas Number : 63314-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07015
Availability details for
Quantity
  • Availability
  • Add to Cart