ENA018110083 Enamine

4-(4-methoxyphenyl)-5-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00834134
  • Cas Number : 63279-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07018
Availability details for
Quantity
  • Availability
  • Add to Cart