ENA018110093 Enamine

1-(2,4,6-trimethylbenzenesulfonyl)piperazine

  • MDL number MFCD03444526
  • Cas Number : 222544-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07031
Availability details for
Quantity
  • Availability
  • Add to Cart